CAS RN 886047-22-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ LY 2119620
Positive allosteric modulator of M2 and M4 receptors
2
Tocris Bioscience™ LY 2119620
Positive allosteric modulator of M2 and M4 receptors