accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNCl

CAS RN 886531-63-1

IUPAC Name: 5-(chloromethyl)-2-phenylpyrimidine
Synonyms: 5-(chloromethyl)-2-phenylpyrimidine
Molecular Weight (g/mol): 204.66
Molecular Formula: C11H9ClN2
InChi Key: VJINEZQXTOUKNJ-UHFFFAOYSA-N
SMILES: ClCC1=CN=C(N=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results