accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COClNH2F

CAS RN 887581-86-4

IUPAC Name: 1-(3-chloro-5-fluoro-4-methoxyphenyl)methanamine
Synonyms: 1-(3-chloro-5-fluoro-4-methoxyphenyl)methanamine
Molecular Weight (g/mol): 189.61
Molecular Formula: C8H9ClFNO
InChi Key: SNAZGTVOOKATFQ-UHFFFAOYSA-N
SMILES: COC1=C(F)C=C(CN)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results