accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O(S)CH3(S)CH3(S)OH(R)CH3CH3OH3COO(R)(S)H3C(R)OH(S)H3C(R)HO(R)OH(S)CH3(R)OH3CCH3(E)(E)(E)(Z)

CAS RN 88899-55-2

IUPAC Name: (3Z,5E,7R,8S,9S,11E,13E,15S,16R)-16-[(2S,3R,4S)-4-[(2R,4R,5S,6R)-2,4-dihydroxy-5-methyl-6-(propan-2-yl)oxan-2-yl]-3-hydroxypentan-2-yl]-8-hydroxy-3,15-dimethoxy-5,7,9,11-tetramethyl-1-oxacyclohexadeca-3,5,11,13-tetraen-2-one
Synonyms: bafilomycin A1 bafilomycin
Molecular Weight (g/mol): 622.84
Molecular Formula: C35H58O9
InChi Key: XDHNQDDQEHDUTM-JQWOJBOSSA-N
SMILES: CO[C@H]1\C=C\C=C(C)\C[C@H](C)[C@H](O)[C@H](C)\C=C(/C)\C=C(OC)\C(=O)O[C@@H]1[C@@H](C)[C@@H](O)[C@H](C)[C@@]1(O)C[C@@H](O)[C@H](C)[C@H](O1)C(C)C
Physical Form 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Type 
  • (3)
Color 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)