CAS RN 88903-69-9
IUPAC Name:
9,30-dioxa-11,25-diazapentacyclo[20.6.2.2⁸,¹¹.0¹⁰,¹⁵.0²⁵,²⁹]dotriacontane
Synonyms:
9,30-dioxa-11,25-diazapentacyclo[20.6.2.2⁸,¹¹.0¹⁰,¹⁵.0²⁵,²⁹]dotriacontane
Molecular Weight (g/mol):
446.72
Molecular Formula:
C28H50N2O2
InChi Key:
PQYOPBRFUUEHRC-UHFFFAOYNA-N
SMILES:
C1CC2CCCCCCC3CCN4CCCC(CCCCCCC5CCN(C1)C2O5)C4O3
1
–
1
of
1
results
1
Tocris Bioscience™ (-)-Xestospongin C
CAS: 88903-69-9 Molecular Formula: C28H50N2O2