accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 89-87-2

CH3H3CNOO

CAS RN 89-87-2

IUPAC Name: 2,4-dimethyl-1-nitrobenzene
Synonyms: 4-nitro-M-xylene
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: BBUPBICWUURTNP-UHFFFAOYSA-N
SMILES: CC1=CC=C(C(C)=C1)[N+]([O-])=O
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)