accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H2N

CAS RN 89-93-0

IUPAC Name: 1-(2-methylphenyl)methanamine
Synonyms: benzenemethanamine, 2-methyl-
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: CJAAPVQEZPAQNI-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1CN
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (4)