Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Methylbenzylamine, MP Biomedicals

Catalog No. ICN206522 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN206522 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN206522 Supplier MP Biomedicals, Inc Supplier No. 0520652205

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 89-93-0
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
InChI Key CJAAPVQEZPAQNI-UHFFFAOYSA-N
Synonym 2-methylbenzylamine, o-tolylmethanamine, o-methylbenzylamine, benzenemethanamine, 2-methyl, o-xylylamine, benzylamine, o-methyl, 2-methylphenyl methanamine, alpha-amino-o-xylene, alpha-amino-2-xylene, 1-2-methylphenyl methanamine
PubChem CID 6993
IUPAC Name (2-methylphenyl)methanamine
SMILES CC1=CC=CC=C1CN

Specifications

Quantity 10 g
Formula Weight 121.18
Chemical Name or Material o-Methylbenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.