accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOSHCl

CAS RN 89025-51-4

IUPAC Name: 2-oxo-2H-4λ⁵-[1,3,4]oxathiazolo[4,5-a]pyridin-4-ylium hydrochloride
Synonyms: 2-oxo-4λ⁵-[1,3,4]oxathiazolo[4,5-a]pyridin-4-ylium hydrochloride
Molecular Weight (g/mol): 190.62
Molecular Formula: C6H5ClNO2S
InChi Key: PVTYXZXONFHZCV-UHFFFAOYSA-N
SMILES: Cl.O=C1OC2=CC=CC=[N+]2S1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results