accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOBrBr

CAS RN 89088-95-9

IUPAC Name: 2,5-dibromo-1λ⁶-thiophene-1,1-dione
Synonyms: 2,5-dibromo-1λ⁶-thiophene-1,1-dione
Molecular Weight (g/mol): 273.93
Molecular Formula: C4H2Br2O2S
InChi Key: LKJJLFANVUXGGB-UHFFFAOYSA-N
SMILES: BrC1=CC=C(Br)S1(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results