accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHNOOOCH3

CAS RN 89464-63-1

IUPAC Name: methyl 2-(2-methoxy-2-oxoacetamido)acetate
Synonyms: methyl 2-(2-methoxy-2-oxoacetamido)acetate
Molecular Weight (g/mol): 175.14
Molecular Formula: C6H9NO5
InChi Key: BNJOZDZCRHCODO-UHFFFAOYSA-N
SMILES: COC(=O)CNC(=O)C(=O)OC
Quantity 
  • (1)
  • (1)
  • (1)
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results