CAS RN 896705-16-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ BMH 21
RNA polymerase I inhibitor; also p53 pathway activator