accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOSNONNS

CAS RN 896795-60-1

IUPAC Name: ethyl 2-({6-oxo-5-[3-(pyrrolidin-1-yl)propyl]-8-thia-3,5-diazatricyclo[7.4.0.0²,⁷]trideca-1(13),2(7),3,9,11-pentaen-4-yl}sulfanyl)acetate
Synonyms: ethyl 2-({6-oxo-5-[3-(pyrrolidin-1-yl)propyl]-8-thia-3,5-diazatricyclo[7.4.0.0²,⁷]trideca-1(13),2(7),3,9,11-pentaen-4-yl}sulfanyl)acetate
Molecular Weight (g/mol): 431.57
Molecular Formula: C21H25N3O3S2
InChi Key: SKDRHRAYBYQVNU-UHFFFAOYSA-N
SMILES: CCOC(=O)CSC1=NC2=C(SC3=CC=CC=C23)C(=O)N1CCCN1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results