accessibility menu, dialog, popup

UserName

Viewing profile as user:
O(E)(E)

CAS RN 897-78-9

IUPAC Name: (2E,6E)-2,6-bis(phenylmethylidene)cyclohexan-1-one
Synonyms: (2E,6E)-2,6-bis(phenylmethylidene)cyclohexan-1-one
Molecular Weight (g/mol): 274.36
Molecular Formula: C20H18O
InChi Key: CTKKGXDAWIAYSA-JSAVKQRWSA-N
SMILES: O=C1\C(CCC\C1=C/C1=CC=CC=C1)=C\C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results