accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 89814-62-0

IUPAC Name: 2,5-bis(thiophen-2-yl)-1H-pyrrole
Synonyms: 2,5-bis(thiophen-2-yl)-1H-pyrrole
Molecular Weight (g/mol): 231.33
Molecular Formula: C12H9NS2
InChi Key: REHRCHHNCOTPBV-UHFFFAOYSA-N
SMILES: N1C(=CC=C1C1=CC=CS1)C1=CC=CS1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results