accessibility menu, dialog, popup

UserName

Viewing profile as user:
O

CAS RN 90-44-8

IUPAC Name: 9,10-dihydroanthracen-9-one
Synonyms: anthrone carbothrone Az-O
Molecular Weight (g/mol): 194.23
Molecular Formula: C14H10O
InChi Key: RJGDLRCDCYRQOQ-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2CC2=CC=CC=C12
Quantity 
  • (1)
  • (7)
  • (8)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (14)
  • (2)
Grade 
  • (7)
  • (2)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (4)
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (3)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (13)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (10)
  • (1)
  • (7)
  • (8)
  • (1)
  • (2)
  • (14)
  • (2)
  • (7)
  • (2)
  • (3)
  • (3)
  • (2)
  • (4)
  • (3)
  • (2)
  • (1)
  • (3)
  • (5)
  • (5)
  • (2)
  • (3)