Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anthrone, Sigma-Aldrich

Catalog No. 01258957 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-957 25 g
01-258-951 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-957 Supplier MilliporeSigma Supelco Supplier No. 31989925G
Add to Cart
Add to Cart

Anthrone, also known as 9(10H)-Anthracenone, is a planar tricyclic aromatic ketone. In acidic conditions, anthrone reacts with carbohydrate to yield a blue-green color.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-44-8
Molecular Weight (g/mol) 194.23 g/mol

Specifications

Color White to Yellow
Quantity 25 g
Percent Purity 97%
Grade ACS
Chemical Name or Material Anthrone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.