Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anthrone, MP Biomedicals™

Catalog No. ICN10069180 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10069180 100 g
ICN10069125 25 g
ICN10069110 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10069180 Supplier MP Biomedicals, Inc Supplier No. 0210069180
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 90-44-8
Molecular Formula C14H10O
Molecular Weight (g/mol) 194.23
MDL Number MFCD00001187
InChI Key RJGDLRCDCYRQOQ-UHFFFAOYSA-N
Synonym 9-10-dihydro-9-oxoanthracene, 9(10H)-Anthracenone
PubChem CID 7018
ChEBI CHEBI:33835
IUPAC Name 9,10-dihydroanthracen-9-one
SMILES O=C1C2=CC=CC=C2CC2=CC=CC=C12

Specifications

Formula Weight 194.22
Color Yellow
Quantity 100 g
Physical Form Powder
Chemical Name or Material Anthrone
storageNote1 Store under nitrogen
Recommended Storage Store at 4°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.