accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOClCl

CAS RN 90-97-1

IUPAC Name: bis(4-chlorophenyl)methanol
Synonyms: bis(4-chlorophenyl)methanol
Molecular Weight (g/mol): 253.12
Molecular Formula: C13H10Cl2O
InChi Key: PHUYGURFBULKPA-UHFFFAOYSA-N
SMILES: OC(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)