Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dichlorobenzhydrol, 98%, Thermo Scientific™

Catalog No. AAL0232722
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0232722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0232722 Supplier Thermo Scientific Chemicals Supplier No. L0232722

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4,4′-Dichlorobenzhydrol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-97-1
Molecular Formula C13H10Cl2O
Molecular Weight (g/mol) 253.12
MDL Number MFCD00000629
InChI Key PHUYGURFBULKPA-UHFFFAOYSA-N
Synonym 4,4'-dichlorobenzhydrol, bis 4-chlorophenyl methanol, p,p'-dichlorobenzhydrol, unii-cr5kh591g4, benzenemethanol, 4-chloro-.alpha.-4-chlorophenyl, bis 4-chlorophenyl methan-1-ol, 4,4'-dichloro-a-phenylbenzylic alcohol, bis 4-chlorphenyl methanol, 4,4'-dichloro-alpha-phenylbenzylic alcohol, 4,4-dicholobenzhydrol
PubChem CID 7033
IUPAC Name bis(4-chlorophenyl)methanol
SMILES OC(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1

Specifications

Melting Point 91°C to 93°C
Quantity 100 g
Beilstein 1878717
Solubility Information Insoluble in water.
Formula Weight 253.13
Percent Purity 98%
Chemical Name or Material 4,4'-Dichlorobenzhydrol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 202-029-6
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.