Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dichlorobenzhydrol 98.0+%, TCI America™
SDP

Catalog No. D191525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D191525G 25 g
D1915250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D191525G Supplier TCI America Supplier No. D191525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 90-97-1
Molecular Formula C13H10Cl2O
Molecular Weight (g/mol) 253.12
MDL Number MFCD00000629
InChI Key PHUYGURFBULKPA-UHFFFAOYSA-N
Synonym 4,4'-dichlorobenzhydrol, bis 4-chlorophenyl methanol, p,p'-dichlorobenzhydrol, unii-cr5kh591g4, benzenemethanol, 4-chloro-.alpha.-4-chlorophenyl, bis 4-chlorophenyl methan-1-ol, 4,4'-dichloro-a-phenylbenzylic alcohol, bis 4-chlorphenyl methanol, 4,4'-dichloro-alpha-phenylbenzylic alcohol, 4,4-dicholobenzhydrol
PubChem CID 7033
IUPAC Name bis(4-chlorophenyl)methanol
SMILES OC(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1

Specifications

Melting Point 93°C
Color White-Yellow
Quantity 25 g
Formula Weight 253.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dichlorobenzhydrol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.