CAS RN 9000-97-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Glutamic oxalacetic transaminase, porcine heart, Thermo Scientific Chemicals
CAS: 9000-97-9 MDL Number: MFCD00131191 Synonym: L-Aspartate: 2-oxoglutarate aminotransferase,Aspartate aminotransferase