accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CBOHHO

CAS RN 90002-36-1

IUPAC Name: (2-ethylphenyl)boronic acid
Synonyms: 2-ethylphenylboronic acid
Molecular Weight (g/mol): 149.98
Molecular Formula: C8H11BO2
InChi Key: QSSPYZOSTJDTTL-UHFFFAOYSA-N
SMILES: CCC1=CC=CC=C1B(O)O
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)