Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. E08185G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0818-5G 5 g
E08181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0818-5G Supplier TCI America Supplier No. E08185G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 90002-36-1
Molecular Formula C8H11BO2
Molecular Weight (g/mol) 149.98
MDL Number MFCD02093075
InChI Key QSSPYZOSTJDTTL-UHFFFAOYSA-N
PubChem CID 4100862
IUPAC Name (2-ethylphenyl)boronic acid
SMILES CCC1=CC=CC=C1B(O)O

Specifications

Melting Point 108°C
Color White-Yellow
Quantity 5 g
Synonym 2-ethylbenzeneboronic acid, 2-ethylphenyl boronic acid, 2-ethylphenyl boranediol, boronic acid, 2-ethylphenyl, 2-ethylphenylboronicacid
Formula Weight 149.98
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.