CAS RN 9001-78-9
IUPAC Name:
2-{[1-(2-{[1-(2-aminopropanoyl)pyrrolidin-2-yl]formamido}acetyl)pyrrolidin-2-yl]formamido}-5-[(diaminomethylidene)amino]pentanoic acid
Synonyms:
alkaline phosphatase
Molecular Weight (g/mol):
496.57
Molecular Formula:
C21H36N8O6
InChi Key:
ITZMJCSORYKOSI-UHFFFAOYNA-N
SMILES:
CC(N)C(=O)N1CCCC1C(=O)NCC(=O)N1CCCC1C(=O)NC(CCCN=C(N)N)C(O)=O
1
–
3
of
3
results
2
Alkaline phosphatase from E. coli, MP Biomedicals™
CAS: 9001-78-9 Molecular Formula: C21H36N8O6 Molecular Weight (g/mol): 496.57 MDL Number: MFCD00131849 InChI Key: ITZMJCSORYKOSI-UHFFFAOYNA-N Synonym: Orthophosphoric Monoester Phosphohydrolase