Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alkaline phosphatase from E. coli, MP Biomedicals™

Catalog No. MP219981780 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kU
100 U
200 U
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219981780 100 U
MP219981782 200 U
MP219981791 1 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP219981780 Supplier MP Biomedicals, Inc Supplier No. 0219981780
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alkaline phosphatase from E. coli ≥30 u/mg protein , Activity- ∽1000 u/mL, Suspension

  • It is used for conjugation to antibodies and other proteins for ELISA, Western blotting, and histochemical detection.
  • It may be used for protein labeling when high sensitivity is required.
  • Alkaline phosphatase is a hydrolase enzyme responsible for removing phosphate groups from many types of molecules, including nucleotides, proteins, and alkaloids.
  • It is most effective in an alkaline environment. It is sometimes used synonymously as basic phosphatase.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9001-78-9
Molecular Formula C21H36N8O6
Molecular Weight (g/mol) 496.57
MDL Number MFCD00131849
InChI Key ITZMJCSORYKOSI-UHFFFAOYNA-N
Synonym Orthophosphoric Monoester Phosphohydrolase

Specifications

Quantity 100 U
Grade Not Available
Physical Form Suspension
storageNote1 Store at +4°C, do not freeze.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.