accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHCH3H3CH3CH3CCH310

CAS RN 9002-93-1

IUPAC Name: 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
Synonyms: triton X polyoxyethylene (11) octyl phenyl ether polyoxyethylene (9) octylphenyl ether hydrol SW poletoxol peg-12 octyl phenyl ether triton X-102 ortho-gynol octoxynol octylphenoxypolyethoxyethanol
Molecular Weight (g/mol): —
Molecular Formula: —
InChi Key: JYCQQPHGFMYQCF-UHFFFAOYSA-N
SMILES: CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
Quantity 
  • (4)
  • (1)
  • (7)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (11)
Molecular Weight (g/mol) 
  • (3)
  • (34)
  • (2)
Grade 
  • (2)
  • (4)
  • (7)
  • (4)
pH 
  • (4)
  • (2)
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (16)
  • (4)
  • (2)
  • (9)
  • (2)
  • (6)
  • (1)
  • (3)
  • (4)
  • (4)
  • (2)
  • (2)
  • (10)
  • (3)
  • (2)
  • (4)
  • (1)
  • (7)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (11)
  • (3)
  • (34)
  • (2)
  • (2)
  • (4)
  • (7)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (3)
  • (1)
  • (1)