Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ 10X Strep-Diluent
SDP

Catalog No. 467109604 Shop All Novus Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-710-9604 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 46-710-9604 Supplier R&D Systems™ Supplier No. 467109604
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

R&D Systems™ 10X Strep-Diluent is a solution that is used as a diluent for the Strep-HRP.

  • Hazard Statements (GHS-US):
    • H319 - Causes serious eye irritation
  • Precautionary Statements (GHS-US):
    • P264 - Wash hands, forearms, and other exposed areas thoroughly after handling.
    • P280 - Wear protective gloves, protective clothing, and eye protection.
    • P305+P351+P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    • P337+P313 - If eye irritation persists: Get medical advice/attention.
  • Store in a dry, cool and well-ventilated place.
  • Keep container closed when not in use.
  • Keep/Store away from direct sunlight, extremely high or low temperatures and incompatible materials.
  • WHMIS Classification: Class D Division 2 Subdivision B - Toxic material causing other toxic effects

Specifications

Quantity 2 mL
CAS 9002-93-1
Content And Storage Store the unopened product at 2°C to 8°C
Product Line R&D Systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.