accessibility menu, dialog, popup

UserName

Viewing profile as user:
∗∗NOn

CAS RN 9003-39-8

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C6H9NO)n
InChi Key: —
SMILES: *-CC(-*)N1CCCC1=O
Quantity 
  • (11)
  • (19)
  • (1)
  • (3)
  • (4)
  • (6)
  • (4)
  • (1)
  • (2)
  • (7)
  • (1)
  • (3)
  • (24)
  • (1)
Molecular Weight (g/mol) 
  • (4)
  • (59)
  • (3)
  • (3)
  • (4)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (9)
  • (18)
Grade 
  • (1)
  • (4)
  • (5)
  • (11)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (51)
  • (23)
  • (2)
  • (5)
  • (7)
  • (21)
  • (23)
  • (6)
  • (28)
  • (15)
  • (11)
  • (19)
  • (1)
  • (3)
  • (4)
  • (6)
  • (4)
  • (1)
  • (2)
  • (7)
  • (1)
  • (3)
  • (24)
  • (1)
  • (4)
  • (59)
  • (3)
  • (3)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (9)
  • (18)
  • (1)
  • (4)
  • (5)
  • (11)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (4)