Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyvinylpyrrolidone K-15, Spectrum™ Chemical
SDP

Catalog No. 18605414 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-414 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-605-414 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PO146100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PO146, 9003-39-8, (C6H9NO)n

Spectrum™ Chemical Polyvinylpyrrolidone K-15 is also known as polyvidone or povidone. It has been used for trauma victims as a plasma volume expander, as well as a binder in some pharmaceutical tablets. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 9003-39-8
Molecular Formula (C6H9NO)n
Molecular Weight (g/mol) 111.14
MDL Number MFCD01076626
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES *-CC(-*)N1CCCC1=O

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 100 g
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.