CAS RN 9003-98-9
IUPAC Name:
Deoxyribonuclease
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
*
1
–
4
of
4
results
1
Deoxyribonuclease I, MP Biomedicals™
CAS: 9003-98-9
2
Deoxyribonuclease I from Bovine pancreas, MP Biomedicals™
Deoxyribonuclease I from Bovine pancreas 50,000 Dornase u/mg solid, White powder
3
MilliporeSigma™ Calbiochem™ DNase I, Bovine Pancreas
Acts on single and double stranded DNA
4
Promotions Available
DNase I, Spectrum™ Chemical