Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deoxyribonuclease I from Bovine pancreas, MP Biomedicals™

Catalog No. MP21900625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
0.5 mU
1 mU
10 mU
250 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21900625 0.5 mU
ICN10057583 250 mg
ICN19006210 10 mU
MP219006201 1 mU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP21900625 Supplier MP Biomedicals, Inc Supplier No. 02190062.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deoxyribonuclease I from Bovine pancreas 50,000 Dornase u/mg solid, White powder

  • Deoxyribonuclease from beef pancreas, DNase I, was first crystallized by Kunitz.
  • It is an endonuclease which splits phosphodiester linkages, preferentially adjacent to a pyrimidine nucleotide yielding 5'-phosphate terminated polynucleotides with a free hydroxyl group on position 3'.
  • The average chain of limit digest is a tetranucleotide. DNase I acts upon single chain DNA, and upon double-stranded DNA and chromatin.
  • In the latter case, although histones restrict susceptibility to nuclease action, over a period of time nearly all chromatin DNA is acted upon.
  • According to Mirsky and Silverman, this could result from the looseness of histone attachment to DNA.
  • They found that lysine-rich histones more effectively block DNase access to DNA than arginine-rich histones
TRUSTED_SUSTAINABILITY

Specifications

Color White
pH 7.8 optimum (literature)
CAS 9003-98-9
Content And Storage Store at -0°C.
Quantity 0.5 mU
Synonym Deoxyribonucleate 5′-oligonucleotido-hydrolase
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.