CAS RN 9004-57-3
Thermo Scientific Chemicals Ethyl cellulose, ethoxyl content 48%, 300 cps
CAS: 9004-57-3 Molecular Formula: C23H24N6O4 Molecular Weight (g/mol): 448.483 MDL Number: MFCD00131037 InChI Key: ARSKJXYLLONUAJ-UHFFFAOYSA-N Synonym: n-butyl-n-2-acetoxyethyl-4-4-nitro-2,6-dicyanophenyl azo-3-methylbenzeneamine,1,3-benzenedicarbonitrile, 2-4-2-acetyloxy ethyl butylamino-2-methylphenyl azo-5-nitro,1,3-benzenedicarbonitrile, 2-2-4-2-acetyloxy ethyl butylamino-2-methylphenyl diazenyl-5-nitro,azo dye n5,4-2,6-dicyano-4-nitrophenyl azo-3-methylphenyl amino hexyl acetate,ethyl cellulose, viscosity ethoxyl,2-n-butyl-4-2,6-dicyano-4-nitrophenyl diazenyl-3-methylanilino ethyl acetate,4-n-2-acetoxyethyl butyl amino-2',6'-dicyano-2-methyl-4'-nitroazobenzene PubChem CID: 166304 IUPAC Name: 2-[N-butyl-4-[(2,6-dicyano-4-nitrophenyl)diazenyl]-3-methylanilino]ethyl acetate SMILES: CCCCN(CCOC(=O)C)C1=CC(=C(C=C1)N=NC2=C(C=C(C=C2C#N)[N+](=O)[O-])C#N)C
Thermo Scientific Chemicals Ethyl cellulose, ethoxyl content 48%, 22 cps
CAS: 9004-57-3 Molecular Formula: C23H24N6O4 Molecular Weight (g/mol): 448.483 MDL Number: MFCD00131037 InChI Key: ARSKJXYLLONUAJ-UHFFFAOYSA-N Synonym: n-butyl-n-2-acetoxyethyl-4-4-nitro-2,6-dicyanophenyl azo-3-methylbenzeneamine,1,3-benzenedicarbonitrile, 2-4-2-acetyloxy ethyl butylamino-2-methylphenyl azo-5-nitro,1,3-benzenedicarbonitrile, 2-2-4-2-acetyloxy ethyl butylamino-2-methylphenyl diazenyl-5-nitro,azo dye n5,4-2,6-dicyano-4-nitrophenyl azo-3-methylphenyl amino hexyl acetate,ethyl cellulose, viscosity ethoxyl,2-n-butyl-4-2,6-dicyano-4-nitrophenyl diazenyl-3-methylanilino ethyl acetate,4-n-2-acetoxyethyl butyl amino-2',6'-dicyano-2-methyl-4'-nitroazobenzene PubChem CID: 166304 IUPAC Name: 2-[N-butyl-4-[(2,6-dicyano-4-nitrophenyl)diazenyl]-3-methylanilino]ethyl acetate SMILES: CCCCN(CCOC(=O)C)C1=CC(=C(C=C1)N=NC2=C(C=C(C=C2C#N)[N+](=O)[O-])C#N)C
Ethylcellulose, 10 mPa.s, NF, 44-51%, Spectrum™ Chemical
CAS: 9004-57-3 Molecular Formula: C23H24N6O4 Molecular Weight (g/mol): 448.48 InChI Key: ARSKJXYLLONUAJ-UHFFFAOYSA-N IUPAC Name: 2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate SMILES: CCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N
Ethyl Cellulose, FCC, 44-50%, Spectrum™ Chemical
CAS: 9004-57-3 Molecular Formula: C23H24N6O4 Molecular Weight (g/mol): 448.48 InChI Key: ARSKJXYLLONUAJ-UHFFFAOYSA-N IUPAC Name: 2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate SMILES: CCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N
Thermo Scientific Chemicals Ethyl cellulose
CAS: 9004-57-3 Molecular Formula: C23H24N6O4 Molecular Weight (g/mol): 448.483 MDL Number: MFCD00131037 InChI Key: ARSKJXYLLONUAJ-UHFFFAOYSA-N Synonym: n-butyl-n-2-acetoxyethyl-4-4-nitro-2,6-dicyanophenyl azo-3-methylbenzeneamine,1,3-benzenedicarbonitrile, 2-4-2-acetyloxy ethyl butylamino-2-methylphenyl azo-5-nitro,1,3-benzenedicarbonitrile, 2-2-4-2-acetyloxy ethyl butylamino-2-methylphenyl diazenyl-5-nitro,azo dye n5,4-2,6-dicyano-4-nitrophenyl azo-3-methylphenyl amino hexyl acetate,ethyl cellulose, viscosity ethoxyl,2-n-butyl-4-2,6-dicyano-4-nitrophenyl diazenyl-3-methylanilino ethyl acetate,4-n-2-acetoxyethyl butyl amino-2',6'-dicyano-2-methyl-4'-nitroazobenzene PubChem CID: 166304 IUPAC Name: 2-[N-butyl-4-[(2,6-dicyano-4-nitrophenyl)diazenyl]-3-methylanilino]ethyl acetate SMILES: CCCCN(CCOC(=O)C)C1=CC(=C(C=C1)N=NC2=C(C=C(C=C2C#N)[N+](=O)[O-])C#N)C
Thermo Scientific Chemicals Ethyl cellulose, ethoxyl content 48%, 10 cps
CAS: 9004-57-3 Molecular Formula: C23H24N6O4 Molecular Weight (g/mol): 448.483 MDL Number: MFCD00131037 InChI Key: ARSKJXYLLONUAJ-UHFFFAOYSA-N Synonym: n-butyl-n-2-acetoxyethyl-4-4-nitro-2,6-dicyanophenyl azo-3-methylbenzeneamine,1,3-benzenedicarbonitrile, 2-4-2-acetyloxy ethyl butylamino-2-methylphenyl azo-5-nitro,1,3-benzenedicarbonitrile, 2-2-4-2-acetyloxy ethyl butylamino-2-methylphenyl diazenyl-5-nitro,azo dye n5,4-2,6-dicyano-4-nitrophenyl azo-3-methylphenyl amino hexyl acetate,ethyl cellulose, viscosity ethoxyl,2-n-butyl-4-2,6-dicyano-4-nitrophenyl diazenyl-3-methylanilino ethyl acetate,4-n-2-acetoxyethyl butyl amino-2',6'-dicyano-2-methyl-4'-nitroazobenzene PubChem CID: 166304 IUPAC Name: 2-[N-butyl-4-[(2,6-dicyano-4-nitrophenyl)diazenyl]-3-methylanilino]ethyl acetate SMILES: CCCCN(CCOC(=O)C)C1=CC(=C(C=C1)N=NC2=C(C=C(C=C2C#N)[N+](=O)[O-])C#N)C
Ethyl Cellulose, 9-11MPA.S, 5% in Toluene + Ethanol (80:20) at 25°C, TCI America™
CAS: 9004-57-3 Molecular Formula: C23H24N6O4 Molecular Weight (g/mol): 448.483 MDL Number: MFCD00131037 InChI Key: ARSKJXYLLONUAJ-UHFFFAOYSA-N Synonym: n-butyl-n-2-acetoxyethyl-4-4-nitro-2,6-dicyanophenyl azo-3-methylbenzeneamine,1,3-benzenedicarbonitrile, 2-4-2-acetyloxy ethyl butylamino-2-methylphenyl azo-5-nitro,1,3-benzenedicarbonitrile, 2-2-4-2-acetyloxy ethyl butylamino-2-methylphenyl diazenyl-5-nitro,azo dye n5,4-2,6-dicyano-4-nitrophenyl azo-3-methylphenyl amino hexyl acetate,ethyl cellulose, viscosity ethoxyl,2-n-butyl-4-2,6-dicyano-4-nitrophenyl diazenyl-3-methylanilino ethyl acetate,4-n-2-acetoxyethyl butyl amino-2',6'-dicyano-2-methyl-4'-nitroazobenzene PubChem CID: 166304 IUPAC Name: 2-[N-butyl-4-[(2,6-dicyano-4-nitrophenyl)diazenyl]-3-methylanilino]ethyl acetate SMILES: CCCCN(CCOC(=O)C)C1=CC(=C(C=C1)N=NC2=C(C=C(C=C2C#N)[N+](=O)[O-])C#N)C
Ethyl Cellulose, 45-55MPA., 5% in Toluene + Ethanol (80:20) at 25°C, TCI America™
CAS: 9004-57-3 Molecular Formula: C23H24N6O4 Molecular Weight (g/mol): 448.483 MDL Number: MFCD00131037 InChI Key: ARSKJXYLLONUAJ-UHFFFAOYSA-N Synonym: n-butyl-n-2-acetoxyethyl-4-4-nitro-2,6-dicyanophenyl azo-3-methylbenzeneamine,1,3-benzenedicarbonitrile, 2-4-2-acetyloxy ethyl butylamino-2-methylphenyl azo-5-nitro,1,3-benzenedicarbonitrile, 2-2-4-2-acetyloxy ethyl butylamino-2-methylphenyl diazenyl-5-nitro,azo dye n5,4-2,6-dicyano-4-nitrophenyl azo-3-methylphenyl amino hexyl acetate,ethyl cellulose, viscosity ethoxyl,2-n-butyl-4-2,6-dicyano-4-nitrophenyl diazenyl-3-methylanilino ethyl acetate,4-n-2-acetoxyethyl butyl amino-2',6'-dicyano-2-methyl-4'-nitroazobenzene PubChem CID: 166304 IUPAC Name: 2-[N-butyl-4-[(2,6-dicyano-4-nitrophenyl)diazenyl]-3-methylanilino]ethyl acetate SMILES: CCCCN(CCOC(=O)C)C1=CC(=C(C=C1)N=NC2=C(C=C(C=C2C#N)[N+](=O)[O-])C#N)C
Ethyl Cellulose, 18-22MPA., 5% in Toluene + Ethanol (80:20) at 25°C, TCI America™
CAS: 9004-57-3 Molecular Formula: C23H24N6O4 Molecular Weight (g/mol): 448.483 MDL Number: MFCD00131037 InChI Key: ARSKJXYLLONUAJ-UHFFFAOYSA-N Synonym: n-butyl-n-2-acetoxyethyl-4-4-nitro-2,6-dicyanophenyl azo-3-methylbenzeneamine,1,3-benzenedicarbonitrile, 2-4-2-acetyloxy ethyl butylamino-2-methylphenyl azo-5-nitro,1,3-benzenedicarbonitrile, 2-2-4-2-acetyloxy ethyl butylamino-2-methylphenyl diazenyl-5-nitro,azo dye n5,4-2,6-dicyano-4-nitrophenyl azo-3-methylphenyl amino hexyl acetate,ethyl cellulose, viscosity ethoxyl,2-n-butyl-4-2,6-dicyano-4-nitrophenyl diazenyl-3-methylanilino ethyl acetate,4-n-2-acetoxyethyl butyl amino-2',6'-dicyano-2-methyl-4'-nitroazobenzene PubChem CID: 166304 IUPAC Name: 2-[N-butyl-4-[(2,6-dicyano-4-nitrophenyl)diazenyl]-3-methylanilino]ethyl acetate SMILES: CCCCN(CCOC(=O)C)C1=CC(=C(C=C1)N=NC2=C(C=C(C=C2C#N)[N+](=O)[O-])C#N)C
Ethyl Cellulose, 5% in Toluene + Ethanol (80:20) at 25°C, TCI America™
CAS: 9004-57-3 Molecular Formula: C23H24N6O4 Molecular Weight (g/mol): 448.483 MDL Number: MFCD00131037 InChI Key: ARSKJXYLLONUAJ-UHFFFAOYSA-N Synonym: n-butyl-n-2-acetoxyethyl-4-4-nitro-2,6-dicyanophenyl azo-3-methylbenzeneamine,1,3-benzenedicarbonitrile, 2-4-2-acetyloxy ethyl butylamino-2-methylphenyl azo-5-nitro,1,3-benzenedicarbonitrile, 2-2-4-2-acetyloxy ethyl butylamino-2-methylphenyl diazenyl-5-nitro,azo dye n5,4-2,6-dicyano-4-nitrophenyl azo-3-methylphenyl amino hexyl acetate,ethyl cellulose, viscosity ethoxyl,2-n-butyl-4-2,6-dicyano-4-nitrophenyl diazenyl-3-methylanilino ethyl acetate,4-n-2-acetoxyethyl butyl amino-2',6'-dicyano-2-methyl-4'-nitroazobenzene PubChem CID: 166304 IUPAC Name: 2-[N-butyl-4-[(2,6-dicyano-4-nitrophenyl)diazenyl]-3-methylanilino]ethyl acetate SMILES: CCCCN(CCOC(=O)C)C1=CC(=C(C=C1)N=NC2=C(C=C(C=C2C#N)[N+](=O)[O-])C#N)C
Sigma Aldrich Ethyl cellulose
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Ethyl cellulose
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Ethyl cellulose
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Ethyl cellulose
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Ethyl cellulose
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
- 1
- 2