Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Cellulose, 18-22MPA., 5% in Toluene + Ethanol (80:20) at 25°C, TCI America™
SDP

Catalog No. E007225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E007225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E007225G Supplier TCI America Supplier No. E007225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 9004-57-3
Molecular Formula C23H24N6O4
Molecular Weight (g/mol) 448.483
MDL Number MFCD00131037
InChI Key ARSKJXYLLONUAJ-UHFFFAOYSA-N
Synonym n-butyl-n-2-acetoxyethyl-4-4-nitro-2,6-dicyanophenyl azo-3-methylbenzeneamine, 1,3-benzenedicarbonitrile, 2-4-2-acetyloxy ethyl butylamino-2-methylphenyl azo-5-nitro, 1,3-benzenedicarbonitrile, 2-2-4-2-acetyloxy ethyl butylamino-2-methylphenyl diazenyl-5-nitro, azo dye n5, 4-2,6-dicyano-4-nitrophenyl azo-3-methylphenyl amino hexyl acetate, ethyl cellulose, viscosity ethoxyl, 2-n-butyl-4-2,6-dicyano-4-nitrophenyl diazenyl-3-methylanilino ethyl acetate, 4-n-2-acetoxyethyl butyl amino-2',6'-dicyano-2-methyl-4'-nitroazobenzene
PubChem CID 166304
IUPAC Name 2-[N-butyl-4-[(2,6-dicyano-4-nitrophenyl)diazenyl]-3-methylanilino]ethyl acetate
SMILES CCCCN(CCOC(=O)C)C1=CC(=C(C=C1)N=NC2=C(C=C(C=C2C#N)[N+](=O)[O-])C#N)C

Specifications

Color White-Yellow
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Ethyl Cellulose [18-22mPa.s, 5% in Toluene + Ethanol (80:20) at 25°C]
EINECSNumber (8)-0186
RTECSNumber FJ5950500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.