CAS RN 9004-62-0
Hydroxyethyl Cellulose, 3,400 cps, Spectrum™ Chemical
CAS: 9004-62-0 Molecular Formula: C29H52O21 Molecular Weight (g/mol): 736.71 InChI Key: CWSZBVAUYPTXTG-UHFFFAOYNA-N IUPAC Name: 5-{[6-({[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}methyl)-3,4-dihydroxy-5-{[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}oxan-2-yl]oxy}-6-(hydroxymethyl)-2-methyloxane-3,4-diol SMILES: COC1C(O)C(O)C(OCC2OC(OC3C(O)C(O)C(C)OC3CO)C(O)C(O)C2OC2OC(CO)C(OC)C(O)C2OCCO)OC1CO
Hydroxyethyl Cellulose, 100 cP, NF, Spectrum™ Chemical
CAS: 9004-62-0 Molecular Formula: C29H52O21 Molecular Weight (g/mol): 736.71 InChI Key: CWSZBVAUYPTXTG-UHFFFAOYNA-N IUPAC Name: 5-{[6-({[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}methyl)-3,4-dihydroxy-5-{[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}oxan-2-yl]oxy}-6-(hydroxymethyl)-2-methyloxane-3,4-diol SMILES: COC1C(O)C(O)C(OCC2OC(OC3C(O)C(O)C(C)OC3CO)C(O)C(O)C2OC2OC(CO)C(OC)C(O)C2OCCO)OC1CO
Hydroxyethyl Cellulose, 5,000 cP, NF, Spectrum™ Chemical
CAS: 9004-62-0 Molecular Formula: C29H52O21 Molecular Weight (g/mol): 736.71 InChI Key: CWSZBVAUYPTXTG-UHFFFAOYNA-N IUPAC Name: 5-{[6-({[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}methyl)-3,4-dihydroxy-5-{[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}oxan-2-yl]oxy}-6-(hydroxymethyl)-2-methyloxane-3,4-diol SMILES: COC1C(O)C(O)C(OCC2OC(OC3C(O)C(O)C(C)OC3CO)C(O)C(O)C2OC2OC(CO)C(OC)C(O)C2OCCO)OC1CO
Hydroxyethyl Cellulose (4,500-6,500mPa.s, 2% in Water at 25°C), TCI America™
CAS: 9004-62-0 Molecular Formula: C29H52O21 Molecular Weight (g/mol): 736.714 MDL Number: MFCD00072770 InChI Key: CWSZBVAUYPTXTG-UHFFFAOYSA-N PubChem CID: 24846132 IUPAC Name: 5-[6-[[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxymethyl]-3,4-dihydroxy-5-[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxyoxan-2-yl]oxy-6-(hydroxymethyl)-2-methyloxane-3,4-diol SMILES: CC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)COC3C(C(C(C(O3)CO)OC)O)O)OC4C(C(C(C(O4)CO)OC)O)OCCO)O)O)O)O
Hydroxyethyl Cellulose (200-300mPa.s, 2% in Water at 20°C), TCI America™
CAS: 9004-62-0 Molecular Formula: C29H52O21 Molecular Weight (g/mol): 736.714 MDL Number: MFCD00072770 InChI Key: CWSZBVAUYPTXTG-UHFFFAOYSA-N PubChem CID: 24846132 IUPAC Name: 5-[6-[[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxymethyl]-3,4-dihydroxy-5-[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxyoxan-2-yl]oxy-6-(hydroxymethyl)-2-methyloxane-3,4-diol SMILES: CC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)COC3C(C(C(C(O3)CO)OC)O)O)OC4C(C(C(C(O4)CO)OC)O)OCCO)O)O)O)O
Hydroxyethyl Cellulose (800-1,500mPa.s, 2% in Water at 20°C), TCI America™
CAS: 9004-62-0 Molecular Formula: C29H52O21 Molecular Weight (g/mol): 736.714 MDL Number: MFCD00072770 InChI Key: CWSZBVAUYPTXTG-UHFFFAOYSA-N PubChem CID: 24846132 IUPAC Name: 5-[6-[[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxymethyl]-3,4-dihydroxy-5-[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxyoxan-2-yl]oxy-6-(hydroxymethyl)-2-methyloxane-3,4-diol SMILES: CC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)COC3C(C(C(C(O3)CO)OC)O)O)OC4C(C(C(C(O4)CO)OC)O)OCCO)O)O)O)O
Sigma Aldrich 2-Hydroxyethyl cellulose
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich 2-Hydroxyethyl cellulose
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich 2-Hydroxyethyl cellulose
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich 2-Hydroxyethyl cellulose
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Hydroxyethyl Cellulose, 2,000 cP, NF, Spectrum™ Chemical
CAS: 9004-62-0 Molecular Formula: C29H52O21 Molecular Weight (g/mol): 736.71 InChI Key: CWSZBVAUYPTXTG-UHFFFAOYNA-N IUPAC Name: 5-{[6-({[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}methyl)-3,4-dihydroxy-5-{[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}oxan-2-yl]oxy}-6-(hydroxymethyl)-2-methyloxane-3,4-diol SMILES: COC1C(O)C(O)C(OCC2OC(OC3C(O)C(O)C(C)OC3CO)C(O)C(O)C2OC2OC(CO)C(OC)C(O)C2OCCO)OC1CO