Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxyethyl Cellulose, 100 cP, NF, Spectrum™ Chemical
SDP

Catalog No. 18603297 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-297 45 kg Amber Glass Bottle
18-603-293 100 g Amber Glass Bottle
18-603-294 500 g Amber Glass Bottle
18-603-295 2.5 kg Amber Glass Bottle
18-603-296 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-603-297 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. HY11245KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

HY112, 9004-62-0

Spectrum™ Chemical Hydroxyethyl Cellulose, 100 cP, NF is a non-ionic, water soluble polymer used as a thickening agent and gelling agent for aqueous pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9004-62-0
Molecular Formula C29H52O21
Molecular Weight (g/mol) 736.71
InChI Key CWSZBVAUYPTXTG-UHFFFAOYNA-N
IUPAC Name 5-{[6-({[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}methyl)-3,4-dihydroxy-5-{[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}oxan-2-yl]oxy}-6-(hydroxymethyl)-2-methyloxane-3,4-diol
SMILES COC1C(O)C(O)C(OCC2OC(OC3C(O)C(O)C(C)OC3CO)C(O)C(O)C2OC2OC(CO)C(OC)C(O)C2OCCO)OC1CO

Specifications

CAS Max % 100%
Ignition Residue 5%
Loss on Drying 10%
Packaging Amber Glass Bottle
Quantity 45 kg
Grade NF
Physical Form Powder or Waxy Solid
Chemical Name or Material Hydroxyethyl Cellulose, 100 cP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.