accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOHOHHOHOOHOHOHHOOONOOONOOONOOOOONOOONOOONOOONOOONOO

CAS RN 9004-70-0

IUPAC Name: 2,3-bis(nitrooxy)-6-[(nitrooxy)methyl]-5-{[3,4,5-tris(nitrooxy)-6-[(nitrooxy)methyl]oxan-2-yl]oxy}oxan-4-yl nitrate; 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Synonyms: 2,3-bis(nitrooxy)-6-[(nitrooxy)methyl]-5-{[3,4,5-tris(nitrooxy)-6-[(nitrooxy)methyl]oxan-2-yl]oxy}oxan-4-yl nitrate; starch
Molecular Weight (g/mol): 1044.57
Molecular Formula: C24H36N8O38
InChi Key: OWWGYSXLINVAES-UHFFFAOYNA-N
SMILES: OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O.[O-][N+](=O)OCC1OC(O[N+]([O-])=O)C(O[N+]([O-])=O)C(O[N+]([O-])=O)C1OC1OC(CO[N+]([O-])=O)C(O[N+]([O-])=O)C(O[N+]([O-])=O)C1O[N+]([O-])=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
Grade 
  • (7)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (7)
  • (2)
  • (7)
  • (1)
  • (7)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (7)
  • (3)
  • (3)