accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOOOCH3CH3

CAS RN 9005-08-7

IUPAC Name: 2-(2-{2-[2-(octadecanoyloxy)ethoxy]ethoxy}ethoxy)ethyl octadecanoate
Synonyms: 2-(2-{2-[2-(octadecanoyloxy)ethoxy]ethoxy}ethoxy)ethyl octadecanoate
Molecular Weight (g/mol): 727.17
Molecular Formula: C44H86O7
InChi Key: MQFYRUGXOJAUQK-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)OCCOCCOCCOCCOC(=O)CCCCCCCCCCCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results