accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COClOCH3

CAS RN 90282-99-8

IUPAC Name: 1-chloro-2,3-dimethoxybenzene
Synonyms: 1-chloro-2,3-dimethoxybenzene
Molecular Weight (g/mol): 172.61
Molecular Formula: C8H9ClO2
InChi Key: JVCXXMPHTZGGNV-UHFFFAOYSA-N
SMILES: COC1=CC=CC(Cl)=C1OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results