CAS RN 9045-23-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CERILLIANT™ beta-Lactoglobulin from bovine milk, certified reference material, Sold by MilliporeSigma™ Supelco™
Certified reference material