CAS RN 9054-89-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Superoxide Dismutase, MP Biomedicals™
Catalyzes the dismutation of superoxide into oxygen and hydrogen peroxide. Superoxide Dismutase, MP Biomedicals is an antioxidant enzyme and an important defense in cells exposed to oxygen. It is a low molecular weight metalloprotein present in aerobic ce