accessibility menu, dialog, popup

UserName

Viewing profile as user:
INOOFFFSOOO

CAS RN 905718-45-8

IUPAC Name: (4-nitrophenyl)(phenyl)iodanium trifluoromethanesulfonate
Synonyms: (4-nitrophenyl)(phenyl)iodanium triflate
Molecular Weight (g/mol): 475.18
Molecular Formula: C13H9F3INO5S
InChi Key: OPSOSRFTIUMCJF-UHFFFAOYSA-M
SMILES: [O-]S(=O)(=O)C(F)(F)F.[O-][N+](=O)C1=CC=C([I+]C2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results