accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)CH3(S)H2NON(S)OHN(S)(S)CH3CH3OHO

CAS RN 90614-48-5

IUPAC Name: (2S,3S)-2-{[(2S)-1-[(2S,3S)-2-amino-3-methylpentanoyl]pyrrolidin-2-yl]formamido}-3-methylpentanoic acid
Synonyms: (2S,3S)-2-{[(2S)-1-[(2S,3S)-2-amino-3-methylpentanoyl]pyrrolidin-2-yl]formamido}-3-methylpentanoic acid
Molecular Weight (g/mol): 341.45
Molecular Formula: C17H31N3O4
InChi Key: JNTMAZFVYNDPLB-PEDHHIEDSA-N
SMILES: CC[C@H](C)[C@H](N)C(=O)N1CCC[C@H]1C(=O)N[C@@H]([C@@H](C)CC)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)