accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 90866-33-4

IUPAC Name: ethyl (3R)-4-chloro-3-hydroxybutanoate
Synonyms: ethyl (3R)-4-chloro-3-hydroxybutanoate
Molecular Weight (g/mol): 166.6
Molecular Formula: C6H11ClO3
InChi Key: ZAJNMXDBJKCCAT-RXMQYKEDSA-N
SMILES: CCOC(=O)C[C@@H](O)CCl
Boiling Point 
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)