Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (R)-4-Chloro-3-hydroxybutyrate 97.0+%, TCI America™
SDP

Catalog No. C17335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17335G 5 g
C173325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17335G Supplier TCI America Supplier No. C17335G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 90866-33-4
Molecular Formula C6H11ClO3
Molecular Weight (g/mol) 166.601
MDL Number MFCD00211242
InChI Key ZAJNMXDBJKCCAT-RXMQYKEDSA-N
Synonym ethyl r-+-4-chloro-3-hydroxybutyrate, r-ethyl 4-chloro-3-hydroxybutanoate, ethyl 3r-4-chloro-3-hydroxybutanoate, ethyl r-4-chloro-3-hydroxybutyrate, ethyl r-+-4-chloro-3-hydroxybutanoate, r-4-chloro-3-hydroxybutyric acid ethyl ester, s-4-chloro-3-hydroxy-butyric acid ethyl ester, r-4-chloro-3-hydroxy-n-butyric acid ethyl ester, ethyl r-4-chloro-3-hydroxybutanoate, butanoic acid, 4-chloro-3-hydroxy-, ethyl ester, 3r
PubChem CID 2734445
IUPAC Name ethyl (3R)-4-chloro-3-hydroxybutanoate
SMILES CCOC(=O)CC(CCl)O

Specifications

Color Yellow
Boiling Point 95°C
Quantity 5 g
Formula Weight 166.60
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl (R)-4-Chloro-3-hydroxybutyrate
EINECSNumber (2)-3796
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.