CAS RN 909250-29-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Eg5 Inhibitor IV, VS-83
The Eg5 Inhibitor IV, VS-83, also referenced under CAS 909250-29-9, controls the biological activity of Eg5. This small molecule/inhibitor is primarily used for Cell Structure applications.