accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNH3C

CAS RN 91-55-4

IUPAC Name: 2,3-dimethyl-1H-indole
Synonyms: 1H-indole, 2,3-dimethyl-
Molecular Weight (g/mol): 145.21
Molecular Formula: C10H11N
InChi Key: PYFVEIDRTLBMHG-UHFFFAOYSA-N
SMILES: CC1=C(C)C2=CC=CC=C2N1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (1)
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)