Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylindole, 97%

Catalog No. AAA1850409
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1850409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1850409 Supplier Thermo Scientific Chemicals Supplier No. A1850409

missing translation for 'validMainframeMsg'

CAS: 91-55-4 | C10H11N | 145.205 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-55-4
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.205
MDL Number MFCD00005617
InChI Key PYFVEIDRTLBMHG-UHFFFAOYSA-N
Synonym 2,3-dimethylindole, 1h-indole, 2,3-dimethyl, indole, 2,3-dimethyl, 2,3-dimethyl indole, tfw7o9hwzk, unii-tfw7o9hwzk, 2,3-dimethyl-1h-indol, 2,3-dimethyl-indol, 2,3-dimethyl-indole, pubchem7334
PubChem CID 7053
IUPAC Name 2,3-dimethyl-1H-indole
SMILES CC1=C(NC2=CC=CC=C12)C

Specifications

Melting Point 105°C to 107°C
Boiling Point 285°C (750 mmHg)
Quantity 10 g
Beilstein 116662
Formula Weight 145.21
Percent Purity 97%
Chemical Name or Material 2,3-Dimethylindole
EINECSNumber 202-076-2
RTECSNumber NL7185000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.