accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91-94-1

IUPAC Name: 3,3'-dichloro-[1,1'-biphenyl]-4,4'-diamine
Synonyms: 3,3'-dichlorobenzidine
Molecular Weight (g/mol): 253.13
Molecular Formula: C12H10Cl2N2
InChi Key: HUWXDEQWWKGHRV-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1Cl)C1=CC=C(N)C(Cl)=C1
Solvent or Matrix 
  • (2)
  • (1)
  • (1)
Element 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results