Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Dichlorobenzidine Stock Standard, SPEX CertiPrep™
SDP

Catalog No. 625E Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
625E 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 625E Supplier SPEX CertiPrep Supplier No. 625E

missing translation for 'validMainframeMsg'

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
3,3'-Dichlorobenzidine91-94-12000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 91-94-1 , 67-56-1
Molecular Formula C12H10Cl2N2
Molecular Weight (g/mol) 253.126
InChI Key HUWXDEQWWKGHRV-UHFFFAOYSA-N
PubChem CID 7070
ChEBI CHEBI:82315
IUPAC Name 4-(4-amino-3-chlorophenyl)-2-chloroaniline
SMILES C1=CC(=C(C=C1C2=CC(=C(C=C2)N)Cl)Cl)N

Specifications

Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Min % 0.2, 99.8
CAS Max % 0.2, 99.8
Methods For US EPA Method 625
Analyte or Component Names 3,3'-Dichlorobenzidine
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Element 3,3'-Dichlorobenzidine
Solvent or Matrix Methanol
Quantity 1 mL
Chemical Name or Material 3,3'-Dichlorobenzidine Stock Standard
Show More Show Less
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.